Få mere at vide
Oxalic Acid-(1,2-13C2), TRC
Artikelnummer. 30011678
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011678 10 mg
30011054 25 mg
30011536 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011678 Leverandør Toronto Research Chemicals Leverandørnr. TRCO84512310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62654-02-8
Molekylær formel 13C2H2O4
Molekylvægt (g/mol) 92.02
Synonym Ethanedioic Acid-(1,2-13C2), Aktisal-(1,2-13C2), Aquisal-(1,2-13C2), DeerClean-(1,2-13C2), NSC 132055-(1,2-13C2), NSC 151956-(1,2-13C2), NSC 62774-(1,2-13C2), NSC 76990-(1,2-13C2), Ultraplast Activate S 52-(1,2-13C2);
SMIL O=[13C](O)[13C](O)=O
Tekniske data
Kemisk navn eller materiale Oxalic Acid-(1,2-13C2)
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)/i1+1,2+1
Formel vægt 92.002
Procent renhed >95%