missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Overheating Alarm Function for Drying and Heating Chambers

The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.

5930.00 DKK

Specifications

Description Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
Type Adjustable Overheating Alarm
View More Specs

Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
16434169
View Documents
BINDER™
8012-1299
BINDER Chambers ED 23 Drying and Heating Chambers with Natural Convection, 230/400 V Model
5930.00 DKK
Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
 
16444169
View Documents
BINDER™
8012-1300
BINDER Chambers FD 23 Drying and Heating Chambers with Forced Convection, 230 V Model
5930.00 DKK
Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
 
16454169
View Documents
BINDER™
8012-1301
BINDER Chambers FED 400 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 400 V
5930.00 DKK
Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
 
Description

Description

  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications

Specifications

Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
Adjustable Overheating Alarm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BINDER™ Overheating Alarm Function for Drying and Heating Chambers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.