Få mere at vide
Osilodrostat, TRC
Artikelnummer. 30133290
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133290 10 mg
17944955 5 mg
17961945 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133290 Leverandør Toronto Research Chemicals Leverandørnr. TRCO82050510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 928134-65-0
Molekylær formel C13H10FN3
Molekylvægt (g/mol) 227.24
Synonym 4-[(5R)-6,7-Dihydro-5H-pyrrolo[1,2-c]imidazol-5-yl]-3-fluorobenzonitrile;4-(R)-(6,7-Dihydro-5H-pyrrolo[1,2-c]imidazol-5-yl)-3-fluorobenzonitrile;Isturisa, LCI 699, LCI 699NX
IUPAC navn 4-[(5R)-6,7-dihydro-5H-pyrrolo[1,2-c]imidazol-5-yl]-3-fluorobenzonitrile
SMIL Fc1cc(ccc1[C@H]2CCc3cncn23)C#N
Tekniske data
Kemisk navn eller materiale Osilodrostat
Mængde 10 mg
InChI formel InChI=1S/C13H10FN3/c14-12-5-9(6-15)1-3-11(12)13-4-2-10-7-16-8-17(10)13/h1,3,5,7-8,13H,2,4H2/t13-/m1/s1
Formel vægt 227.0859