missing translation for 'onlineSavingsMsg'
Få mere at vide

Oralith Brilliant Pink R, Technical Grade, TRC

Artikelnummer. 30418967
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418967 500 mg
30427721 250 mg
30416946 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30418967 Leverandør Toronto Research Chemicals Leverandørnr. TRCO670605500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2379-74-0
Molekylær formel C18 H10 Cl2 O2 S2
Molekylvægt (g/mol) 393.31
Synonym 6-Chloro-2-(6-chloro-4-methyl-3-oxobenzo[b]thien-2(3H)-ylidene)-4-methylbenzo[b]thiophen-3(2H)-one, 6,6'-Dichloro-4,4'-dimethyl-[δ2,2'(3H,3'H)-bibenzo[b]thiophene]-3,3'-dione, 11484 Red, 5,5'-Dichloro-3,3'-dimethyl-thioindigo, 6,6'-Dichloro-4,4'-dimethylthioindigo, Ahcovat Pink FFD, Ahcovat Printing Pink FF, Amanthrene Pink FF, Amanthrene Pink FFD, Amanthrene Pink FFWP, C.I. 73360, C.I. Pigment Red 181, C.I. Vat Red 1, Calcoloid Pink FFC, Calcoloid Pink FFD, Calcoloid Pink FFRP, Calcoloid Printing Pink FFE, Calcophyl Red FF, Calophyl Pink ZFF, Chemithrene Brilliant Pink R, Ciba Brilliant Pink FR, Ciba Brilliant Pink R, Ciba Pink FF, D and C Red No. 30, D&C Red 30, D&C Red 30, Daltolite Pink FF, Durindone Pink FF, Durindone Pink FF-FA, Durindone Printing Pink FF, Fast Pink Y, Fenanthren Brilliant Pink R, Fenanthren Pink R Spura, Fenidon Pink R, Helanthrene Brilliant Pink R, Helanthrene Pink R, Helindon Pink CN, Helindon Pink R, Helindone Pink CN, Hostavat Brilliant Pink R, Indanthren Brilliant Pink R, Indanthren Brilliant Pink RB, Indanthren Brilliant Pink RP, Indanthren Brilliant Pink RS, Indanthren Brilliant Rose R, Indanthrene Brilliant Pink R, Indanthrene Pink R, Japan Red 226, Lithosol Fast Pink SVP, Mikethrene Brilliant Pink R, Nihonthrene Brilliant Pink R, Nyanthrene Brilliant Pink R, Oracet Pink RF, Oralith Brilliant Pink R, Palanthrene Brilliant Pink R, Paradone Brilliant Pink R, Permanent Pink, Pigment Red 181, Pink FFT, Red No. 226, Romantrene Brilliant Pink FR, Sandothrene Brilliant Pink R, Sanyo Threne Brillant Pink IR, Solanthrene Brilliant Pink F-R, Solanthrene Brilliant Pink R, Solanthrene Brilliant Pink RF, Sulfanthrene Pink FFD, Thioindigo Brilliant Pink Zh, Thioindigo Brilliant Pink ZhP, Tina Brilliant Pink R, Tyrian Brilliant Pink I-R, Vat Pink FF, Vat Pink R, Vat Printing Pink FF, Vat Red 1
IUPAC navn (2E)-6-chloro-2-(6-chloro-4-methyl-3-oxo-1-benzothiophen-2-ylidene)-4-methyl-1-benzothiophen-3-one
SMIL Cc1cc(Cl)cc2S\C(=C/3\Sc4cc(Cl)cc(C)c4C3=O)\C(=O)c12

Tekniske data

Kemisk navn eller materiale VAT Red 1
Mængde 500 mg
InChI formel InChI=1S/C18H10Cl2O2S2/c1-7-3-9(19)5-11-13(7)15(21)17(23-11)18-16(22)14-8(2)4-10(20)6-12(14)24-18/h3-6H,1-2H3/b18-17+
Formel vægt 391.9499
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.