missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ DNA Polymerase I, Fisher BioReagents™

408.47 DKK - 2220.00 DKK

Specifications

Concentration 10 U/μL
Color Colorless
pH 7.5
For Use With (Application) DNA Polymerase I labels DNA by nick translation in conjunction with DNase I, second-strand cDNA synthesis in conjunction with RNase H, incorporation of modified Nucleotides, Reverse Transcriptases
Storage Buffer OPTIZYME™ DNA Pol I Buffer 10X : Tris HCl 500mM (7.5 pH at 25°C), MgCl2 100mM, DTT 10mM
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12807172
View Documents Promotion Details
Fisher Bioreagents
BP8109-1
500 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12817172
View Documents Promotion Details
Fisher Bioreagents
BP8109-5
2500 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

OPTIZYME™ DNA Pol I is a template-dependent DNA Polymerase that catalyzes 5'->3' DNA synthesis, and exhibits 3'->5'-exonuclease, 5'->3' exonuclease and RNase H activity.

Applications and Features:

  • OPTIZYME DNA Polymerase I labels DNA by nick translation in conjunction with DNase I.
  • Second-strand cDNA synthesis in conjunction with RNase H.
  • Incorporation of modified nucleotides (e.g., biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides).
  • Active in buffers for OPTIZYME restriction enzymes and reverse transcriptases.

Supplied with:
10X OPTIZYME DNA Pol I Buffer: 500mM Tris-HCl (pH 7.5 at 25°C), 100mM MgCl2, 10mM DTT

Source: E. Coli cells with a cloned polA gene for E.Coli

Quality Control:
Endodeoxyribonuclease-free

Specifications

Specifications

10 U/μL
7.5
OPTIZYME™ DNA Pol I Buffer 10X : Tris HCl 500mM (7.5 pH at 25°C), MgCl2 100mM, DTT 10mM
E. coli cells with a cloned polA gene
Liquid
Colorless
DNA Polymerase I labels DNA by nick translation in conjunction with DNase I, second-strand cDNA synthesis in conjunction with RNase H, incorporation of modified Nucleotides, Reverse Transcriptases
-20°C
DNA Polymerase I
Videos
SDS
Documents

Documents

Product Certifications