missing translation for 'onlineSavingsMsg'
Få mere at vide

Octanal, Sigma-Aldrich™

Product Code. 15879350
Change view
Click to view available options
Mængde:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
15879350 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15879350 Supplier Sigma Aldrich Supplier No. 8069010100
 Begrænset vare
Forventet forsendelse: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 124-13-0
Smeltepunkt 12°C to 15°C
Tæthed 0.82 g/cm3 at 20°C
Kogepunkt 167°C to 169°C/1013 hPa
Molekylær formel CH3(CH2)6CHO
MDL nummer MFCD00007029
Mængde 100 mL
Synonym Octanal, Caprylaldehyde, Octaldehyde, Octanal
Molekylvægt (g/mol) 128.21
Grad Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.