missing translation for 'onlineSavingsMsg'
Få mere at vide

NVS-PAK1-1, MedChemExpress

Artikelnummer. 16494356
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494356 100 mg 100mg
16482596 25 mg -
16473058 10 mg -
16429626 50 mg -
16425797 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16494356 Leverandør Medchem Express Leverandørnr. HY100519

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress NVS-PAK1-1 is a potent and selective allosteric PAK1 inhibitor with an IC50 of 5 nM.

NVS-PAK1-1 is a potent and selective allosteric PAK1 inhibitor with an IC50 of 5 nM.

Specifications

Kemisk navn eller materiale NVS-PAK1-1
CAS 1783816-74-9
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C23H25ClF3N5O
Mængde 100 mg
Opløselighedsinformation DMSO : ≥ 125 mg/mL (260.45 mM)
SMIL FC1=CC2=C(C=C1)N(CC(F)F)C3=C(C=C(Cl)C=C3)C(N[C@H]4CCN(C(NC(C)C)=O)C4)=N2
Molekylvægt (g/mol) 479.93
Formel vægt 479.93
Procent renhed 99.91%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.