Få mere at vide
NVP-QAV-572, MedChemExpress
Artikelnummer. 16483087
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16483087 50 mg
16477038 10 mg
16437096 100 mg
16429587 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16483087 Leverandør Medchem Express Leverandørnr. HY16355
MedChemExpress NVP-QAV-572 is a PI3K inhibitor extracted from patent US7998990B2, Compound Example 8, has an IC50 of 10 nM.
NVP-QAV-572 is a PI3K inhibitor extracted from patent US7998990B2, Compound Example 8, has an IC50 of 10 nM.
Tekniske data
Kemisk navn eller materiale NVP-QAV-572
CAS 957209-68-6
Farve White
Renhedsgrad noter Research
Molekylær formel C17H19F2N7O3S2
Mængde 50 mg
Opløselighedsinformation DMSO : 125 mg/mL (265.11 mM; Need ultrasonic)
SMIL O=C(NC1=NC(C)=C(C2=CC=C(S(=O)(C)=O)C(F)=C2)S1)NCCC3=NN(CCF)N=N3
Molekylvægt (g/mol) 471.5
Formel vægt 471.5
Procent renhed 98.05%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease