Learn More
Thermo Scientific™ Nunclon™ Sphera™ 96-Well, Nunclon Sphera-Treated, U-Shaped-Bottom Microplate
BIOPHARMA
Color: Clear; Sterility: Sterile; Lid: With Lid
2110.00 DKK - 9535.00 DKK
Specifications
Material Polystyrene
Volume (Metric) Well 300 ÎĽL
Type Microplate
Color Clear
Surface Treatment Nunclon Sphera-Treated
View More Specs
Products 2
Product Code Brand No. per Pack No. per Case Price Quantity & Availability  
Product Code Brand No. per Pack No. per Case Price Quantity & Availability  
15227905
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
174925
1 Microplate 8 Microplates
2110.00 DKK
Pack of 8
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
15396123
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
174929
5 Microplates 50 Microplates
9535.00 DKK
Pack of 50
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
Description
Description
Make your most valuable work your best work. Thermo Scientific™ Nunclon™ Sphera™ MIcroplates feature a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research. Offered in single or multi-dish formats.
Significant qualities of the Nunclon Sphera culture surface:
Optimizing spheroid formation
  • Allows cells to grow in suspension with virtually no cell attachment
  • Supports many different cell types and their ability to generate spheroids
  • Enables consistent, repeatable cell growth
Growing 3D cancer cultures
  • Minimizes variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumor growth, providing a better model system for studying tumor cell progression and efficacy of anticancer agents in vitro
Promoting formation of embryoid bodies (EB)
  • Demonstrates superior quality for embryoid body formation of pluripotent stem cells with minimal spontaneous differentiation
  • Resultant cells able to differentiate into all three germ layers
Specifications
Specifications
Polystyrene
Microplate
Nunclon Sphera-Treated
96
8 x 12
U
300 ÎĽL
Clear
Sterile
Animal origin-free; nonpyrogenic, endotoxin <0.5 EU/mL; USP biological reactivity class VI; SAL 10-6; ANSI/SBS 1-2004
Nunc™
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nunclon™ Sphera™ 96-Well, Nunclon Sphera-Treated, U-Shaped-Bottom Microplate