Learn More
Macherey-Nagel™ Nucleodur™ PolarTec HPLC Columns
Column Format: Analytical Column; Carbon Load: 17%; Pore Size: 110 Ă…; Column Phase: Reversed Phase
6045.00 DKK - 6505.00 DKK
Specifications
Column Chemistry | Octadecyl |
---|---|
Column Format | Analytical Column |
Crosslinking | Octadecyl |
Flow Rate | 0.5 mL/min |
For Use With | For Phenols and Nitrogen Containing Compounds, Polar Compounds Like Basic Pharmaceuticals, Organic Acids, Pesticides, Amino Acids, Water Soluble Vitamins, Etc. Application |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
12798822
|
Macherey-Nagel™
760478.46 |
4.6 mm |
6505.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
12788822
|
Macherey-Nagel™
760478.40 |
4 mm |
6045.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
Specifications
Octadecyl | |
Octadecyl | |
For Phenols and Nitrogen Containing Compounds, Polar Compounds Like Basic Pharmaceuticals, Organic Acids, Pesticides, Amino Acids, Water Soluble Vitamins, Etc. Application | |
Yes | |
8700 psi (600 bar) | |
Spherical | |
1 to 9 | |
0.9 mL/g | |
Acetonitrile/water | |
60°C | |
C18 with Embedded Polar Group | |
17% | |
Yes | |
Stainless Steel |
Analytical Column | |
0.5 mL/min | |
Hydrophobic | |
150 mm | |
Acetonitrile/water | |
3 ÎĽm | |
110 Ă… | |
1 Ea. | |
340 m²/g | |
L1/L60 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
EC Column |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Macherey-Nagel™ Nucleodur™ PolarTec HPLC Columns