Få mere at vide
Merck Millipore Novagen™ KOD Hot Start DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
3220.00 DKK
Produkter 1
Produktkode Brand Quantity Pris Antal & tilgængelighed  
Produktkode Brand Quantity Pris Antal & tilgængelighed  
12838435
Dokumentvisning
Merck Millipore
71086-3
ONLINE EXCLUSIVE PRICE
4380.00 DKK
3220.00 DKK / Hver
Save 1160.00 DKK
26% Slukket
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse
Beskrivelse
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
Videoer
SDS
Dokumenter
Dokumenter
Produktcertifikater
For Research Use Only