missing translation for 'onlineSavingsMsg'
Få mere at vide

Norflurazon, TRC

Artikelnummer. 30410351
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410351 50 mg
30419469 1 g
30418510 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30410351 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68450050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 27314-13-2
Molekylær formel C12 H9 Cl F3 N3 O
Molekylvægt (g/mol) 303.67
Synonym Evital, 4-chloro-5-(methylamino)-2-(α,α,α-trifluoro-m-tolyl)pyridazin-3(2H)-one
IUPAC navn 4-chloro-5-(methylamino)-2-[3-(trifluoromethyl)phenyl]pyridazin-3-one
SMIL CNC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F

Tekniske data

Kemisk navn eller materiale Norflurazon
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C12H9ClF3N3O/c1-17-9-6-18-19(11(20)10(9)13)8-4-2-3-7(5-8)12(14,15)16/h2-6,17H,1H3
Formel vægt 303.0386
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.