Få mere at vide
Norfluoxetine-d5 Hydrochloride (Phenyl-d5), TRC
Artikelnummer. 30416680
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416680 10 mg
30423329 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416680 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68110310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1188265-34-0
Molekylær formel C16H12D5ClF3NO
Molekylvægt (g/mol) 336.79
Synonym γ-[4-(Trifluoromethyl)phenoxy]benzenepropanamine-d5 Hydrochloride, (+/-)-γ-[4-(Trifluoromethyl)phenoxy]benzenepropanamine-d5 Hydrochloride, Desmethylfluoxetine-d5 Hydrochloride
SMIL NCCC(OC1=CC=C(C(F)(F)F)C=C1)C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H].Cl
Tekniske data
Kemisk navn eller materiale Norfluoxetine-d5 Hydrochloride (Phenyl-d5)
Mængde 10 mg
InChI formel InChI=1S/C16H16F3NO.ClH/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;/h1-9,15H,10-11,20H2;1H/i1D,2D,3D,4D,5D;
Formel vægt 336.1265