Få mere at vide
Nintedanib N-Acetyl Impurity, TRC
Artikelnummer. 30482541
Skift visning
Mængde:
2.5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482541 2.5 mg
30471390 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482541 Leverandør Toronto Research Chemicals Leverandørnr. TRCN2018202.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2130844-77-6
Molekylær formel C33H35N5O5
Molekylvægt (g/mol) 581.66
Synonym 1H-Indole-6-carboxylic acid, 1-acetyl-2,3-dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-, methyl ester
IUPAC navn methyl (3Z)-1-acetyl-3-[[4-[methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino]anilino]-phenylmethylidene]-2-oxoindole-6-carboxylate
SMIL COC(=O)c1ccc2\C(=C(\Nc3ccc(cc3)N(C)C(=O)CN4CCN(C)CC4)/c5ccccc5)\C(=O)N(C(=O)C)c2c1
Tekniske data
Kemisk navn eller materiale Nintedanib N-Acetyl Impurity
Mængde 2.5 mg
InChI formel InChI=1S/C33H35N5O5/c1-22(39)38-28-20-24(33(42)43-4)10-15-27(28)30(32(38)41)31(23-8-6-5-7-9-23)34-25-11-13-26(14-12-25)36(3)29(40)21-37-18-16-35(2)17-19-37/h5-15,20,41H,16-19,21H2,1-4H3
Formel vægt 581.2638