Få mere at vide
Nicorandil, 98+%, Thermo Scientific™
A vasodilator that activates ATP-sensitive potassium channels
Brand: Thermo Scientific Alfa Aesar J60879.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 65141-46-0 | |
| 211.18 | |
| LBHIOVVIQHSOQN-UHFFFAOYSA-N | |
| 47528 | |
| 2-[(pyridin-3-yl)formamido]ethyl nitrate |
| C8H9N3O4 | |
| MFCD00186520 | |
| nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin | |
| CHEBI:31905 | |
| [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1 |
Tekniske data
| Nicorandil | |
| C8H9N3O4 | |
| 1g | |
| nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin | |
| [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1 | |
| 211.18 | |
| CHEBI:31905 | |
| ≥98% |
| 65141-46-0 | |
| MFCD00186520 | |
| 14,6521 | |
| LBHIOVVIQHSOQN-UHFFFAOYSA-N | |
| 2-[(pyridin-3-yl)formamido]ethyl nitrate | |
| 47528 | |
| 211.17 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.