Få mere at vide
Niclosamide Piperazinesalt, TRC
Artikelnummer. 30136848
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136848 100 mg
30135393 50 mg
30008171 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136848 Leverandør Toronto Research Chemicals Leverandørnr. TRCN234040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34892-17-6
Molekylær formel C13H8Cl2N2O4 . C 4H10N2
Molekylvægt (g/mol) 413.26
Synonym Piperazine, Compd. With 5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMIL C1CNCCN1.C1=CC(=C(C=C1[N+](=O)[O-])Cl)NC(=O)C2=C(C=CC(=C2)Cl)O
Tekniske data
Kemisk navn eller materiale Niclosamide Piperazinesalt
Mængde 100 mg
InChI formel InChI=1S/C13H8Cl2N2O4.C4H10N2/c14-7-1-4-12(18)9(5-7)13(19)16-11-3-2-8(17(20)21)6-10(11)15;1-2-6-4-3-5-1/h1-6,18H,(H,16,19);5-6H,1-4H2
Formel vægt 412.07