Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
Brand: Cole-Parmer™ 41507-64
Additional Details : Weight : 0.02000kg
Specifications
Luer Fitting | |
Nickle Plated Brass | |
Harsh chemical environments | |
1/16 in. Hose Barb |
Autoclavable | |
Female luer lock | |
1 Pack |