missing translation for 'onlineSavingsMsg'
Learn More

NG25, MedChemExpress

Product Code. 16489097
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16489097 5 mg
16456518 10 mg
16451519 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489097 Supplier Medchem Express Supplier No. HY15434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NG25 is a potent dual TAK1 and MAP4K2 inhibitor, with IC50s of 149 nM and 21.7 nM, respectively.

NG25 is a potent dual TAK1 and MAP4K2 inhibitor, with IC50s of 149 nM and 21.7 nM, respectively.

Specifications

Kemisk navn eller materiale NG25
CAS 1315355-93-1
Farve White
Renhedsgrad noter Research
Molekylær formel C29H30F3N5O2
Mængde 5 mg
Opløselighedsinformation DMSO : 50 mg/mL (93.01 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C(NC1=CC(C(F)(F)F)=C(CN(CC2)CCN2CC)C=C1)C3=CC(OC4=C5C(NC=C5)=NC=C4)=C(C)C=C3
Molekylvægt (g/mol) 537.58
Formel vægt 537.58
Procent renhed 99.09%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.