missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NFIL3 (Human) Recombinant Protein

Human NFIL3 full-length ORF ( AAH08197, 1 a.a. - 462 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004783-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138341

  • 2665.00 DKK / 10µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

  • Sequence: MQLRKMQTVKKEQASLDASSNVDKMMVLNSALTEVSEDSTTGEELLLSEGSVGKNKSSACRRKREFIPDEKKDAMYWEKRRKNNEAAKRSREKRRLNDLVLENKLIALGEENATLKAELLSLKLKFGLISSTAYAQEIQKLSNSTAVYFQDYQTSKSNVSSFVDEHEPSMVSSSCISVIKHSPQSSLSDVSEVSSVEHTQESSVQGSCRSPENKFQIIKQEPMELESYTREPRDDRGSYTASIYQNYMGNSFSGYSHSPPLLQVNRSSSNSPRTSETDDGVVGKSSDGEDEQQVPKGPIHSPVELKHVHATVVKVPEVNSSALPHKLRIKAKAMQIKVEAFDNEFEATQKLSSPIDMTSKRHFELEKHSAPSMVHSSLTPFSVQVTNIQDWSLKSEHWHQKELSGKTQNSFKTGVVEMKDSGYKVSDPENLYLKQGIANLSAEVVSLKRLIATQPISASDSG
Specifications

Specifications

AAH08197
nuclear factor, interleukin 3 regulated
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NFIL3
GST
4783
Wheat germ expression system
10 μg
E4BP4, IL3BP1, NF-IL3A, NFIL3A
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers