missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NFIL3 (Human) Recombinant Protein
Human NFIL3 full-length ORF ( AAH08197, 1 a.a. - 462 a.a.) recombinant protein with GST-tag at N-terminal.
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH08197 |
---|---|
Gene ID (Entrez) | 4783 |
Name | nuclear factor, interleukin 3 regulated |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138341
|
Abnova™
H00004783-P01.10UG |
10 μg |
2665.00 DKK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148341
|
Abnova™
H00004783-P01.25UG |
25 μg |
4040.00 DKK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MQLRKMQTVKKEQASLDASSNVDKMMVLNSALTEVSEDSTTGEELLLSEGSVGKNKSSACRRKREFIPDEKKDAMYWEKRRKNNEAAKRSREKRRLNDLVLENKLIALGEENATLKAELLSLKLKFGLISSTAYAQEIQKLSNSTAVYFQDYQTSKSNVSSFVDEHEPSMVSSSCISVIKHSPQSSLSDVSEVSSVEHTQESSVQGSCRSPENKFQIIKQEPMELESYTREPRDDRGSYTASIYQNYMGNSFSGYSHSPPLLQVNRSSSNSPRTSETDDGVVGKSSDGEDEQQVPKGPIHSPVELKHVHATVVKVPEVNSSALPHKLRIKAKAMQIKVEAFDNEFEATQKLSSPIDMTSKRHFELEKHSAPSMVHSSLTPFSVQVTNIQDWSLKSEHWHQKELSGKTQNSFKTGVVEMKDSGYKVSDPENLYLKQGIANLSAEVVSLKRLIATQPISASDSG
Specifications
AAH08197 | |
nuclear factor, interleukin 3 regulated | |
125% SDS-PAGE Stained with Coomassie Blue | |
E4BP4, IL3BP1, NF-IL3A, NFIL3A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4783 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NFIL3 | |
GST |