missing translation for 'onlineSavingsMsg'
Få mere at vide

Nesbuvir, MedChemExpress

Artikelnummer. 16498757
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16498757 5 mg -
16492297 50 mg -
16416168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16498757 Leverandør Medchem Express Leverandørnr. HY14775

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Nesbuvir is a nonnucleoside inhibitor of the hepatitis C virus (HCV) nonstructural protein 5B (NS5B) polymerase.

Nesbuvir is a nonnucleoside inhibitor of the hepatitis C virus (HCV) nonstructural protein 5B (NS5B) polymerase.

Specifications

Kemisk navn eller materiale Nesbuvir
CAS 691852-58-1
Farve White
Renhedsgrad noter Research
Molekylær formel C22H23FN2O5S
Mængde 5 mg
Synonym HCV-796
Opløselighedsinformation DMSO : ≥ 50 mg/mL (111.98 mM)
SMIL O=C(C1=C(C2=CC=C(F)C=C2)OC3=CC(N(CCO)S(=O)(C)=O)=C(C4CC4)C=C13)NC
Molekylvægt (g/mol) 446.49
Formel vægt 446.49
Procent renhed 98.52%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.