missing translation for 'onlineSavingsMsg'
Få mere at vide

Neostigmine Methyl Sulfate, TRC

Artikelnummer. 30482894
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482894 100 mg
30475593 1 g
30481107 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30482894 Leverandør Toronto Research Chemicals Leverandørnr. TRCN390020100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 51-60-5
Molekylær formel C12 H19 N2 O2 . C H3 O4 S
Molekylvægt (g/mol) 334.39
Synonym Ammonium, (m-hydroxyphenyl)trimethyl-, methyl sulfate, dimethylcarbamate (8CI), Benzenaminium, 3-[[(dimethylamino)carbonyl]oxy]-N,N,N-trimethyl-, methyl sulfate (9CI), (3-Dimethylcarbamoyloxyphenyl)trimethylammonium methyl sulfate, (3-Hydroxyphenyl)trimethylammonium methyl sulfate dimethylcarbamic ester, (m-Hydroxyphenyl)trimethylammonium methyl sulfate dimethylcarbamate, 3-(Dimethylcarbamoxy)phenyl trimethylammonium methyl sulfate, Eustigmin methyl sulfate, GD 65, Hodostin, Intrastigmina, Kirkstigmine methyl sulfate, Leostigmine methyl sulfate, Metastigmin, NSC 93753, Neoeserine methyl sulfate, Neostigmeth, Neostigmine methosulfate, Neostigmine methyl sulfate, Neostigmine mono(methyl sulfate), Normastigmin, Philostigmin methyl sulfate, Polstigmine, Proserine, Proserine methyl sulfate, Prostigmin methyl sulfate, Prostigmine methyl sulfate, Stiglyn, Stigmanol methyl sulfate, Stigmosan methyl sulfate, Synstigmine, Syntostigmin, Syntostigmin (injection), Vagostigmin, Vagostigmine methyl sulfate
IUPAC navn [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate
SMIL COS(=O)(=O)[O-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

Tekniske data

Kemisk navn eller materiale Neostigmine Methyl Sulfate
Mængde 100 mg
InChI formel InChI=1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1
Formel vægt 334.1199
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.