missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neostigmine methyl sulfate, MedChemExpress
Description
Neostigmine methyl sulfate is a reversible inhibitor of acetylcholinesterase, can not cross the blood-brain barrier.
Specifications
Specifications
| Kemisk navn eller materiale | Neostigmine methyl sulfate |
| CAS | 51-60-5 |
| Farve | Off-White |
| Renhedsgrad noter | Research |
| Molekylær formel | C13H22N2O6S |
| Mængde | 100 mg |
| Opløselighedsinformation | H2O : 100 mg/mL (299.05 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (299.05 mM) |
| SMIL | C[N+](C)(C)C1=CC=CC(OC(N(C)C)=O)=C1.O=S(OC)([O-])=O |
| Molekylvægt (g/mol) | 334.39 |
| Formel vægt | 334.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?