Få mere at vide
(-)-Nefopam Hydrochloride, TRC
Artikelnummer. 30472303
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472303 250 mg
30465015 500 mg
30466879 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30472303 Leverandør Toronto Research Chemicals Leverandørnr. TRCN385750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53625-25-5
Molekylær formel C17H19NO . HCl
Molekylvægt (g/mol) 289.8
Synonym (R)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, (1R)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, R-Nefopam Hydrochloride
IUPAC navn (1R)-5-methyl-1-phenyl-1,3,4,6-tetrahydro-2,5-benzoxazocine;hydrochloride
SMIL Cl.CN1CCO[C@H](c2ccccc2)c3ccccc3C1
Tekniske data
Kemisk navn eller materiale (-)-Nefopam Hydrochloride
Mængde 250 mg
InChI formel InChI=1S/C17H19NO.ClH/c1-18-11-12-19-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18;/h2-10,17H,11-13H2,1H3;1H/t17-;/m1./s1
Formel vægt 289.1233