Få mere at vide
Nefopam-d3 Hydrochloride, TRC
Artikelnummer. 30482247
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482247 1 mg
30469133 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482247 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3894021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346603-30-2
Molekylær formel C17H16D3NO . HCl
Molekylvægt (g/mol) 292.8
Synonym 3,4,5,6-Tetrahydro-5-(methyl-d3)-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, (+/-)-Nefopam-d3 Hydrochloride, Acupan-d3, Ajan-d3, Fenazoxine-d3, Fenazoxine-d3 Hydrochloride, R 738-d3
IUPAC navn 1-phenyl-5-(trideuteriomethyl)-1,3,4,6-tetrahydro-2,5-benzoxazocine;hydrochloride
SMIL Cl.[2H]C([2H])([2H])N1CCOC(c2ccccc2)c3ccccc3C1
Tekniske data
Kemisk navn eller materiale Nefopam-d3 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C17H19NO.ClH/c1-18-11-12-19-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18;/h2-10,17H,11-13H2,1H3;1H/i1D3;
Formel vægt 292.1422