Få mere at vide
NAlpha-Fmoc-D-histidine, TRC
Artikelnummer. 30179346
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179346 1 g
30179311 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179346 Leverandør Toronto Research Chemicals Leverandørnr. TRCF6330051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157355-79-8
Molekylær formel C21H19N3O4
Molekylvægt (g/mol) 377.39
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-histidine
IUPAC navn (((9H-fluoren-9-yl)methoxy)carbonyl)-D-histidine
SMIL O=C(O)[C@H](NC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)CC4=CN=CN4
Tekniske data
Kemisk navn eller materiale Nalpha-Fmoc-D-histidine
InChI formel InChI=1S/C21H19N3O4/c25-20(26)19(9-13-10-22-12-23-13)24-21(27)28-11-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,10,12,18-19H,9,11H2,(H,22,23)(H,24,27)(H,25,26)/t19-/m1/s1
Mængde 1 g
Formel vægt 377.14