Få mere at vide
Nabumetone, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J63072.14
| Mængde | 25g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Anti-inflammatory, anti-bacterial
- A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase
Kemiske identifikatorer
| 42924-53-8 | |
| 228.291 | |
| BLXXJMDCKKHMKV-UHFFFAOYSA-N | |
| 4409 | |
| 4-(6-methoxynaphthalen-2-yl)butan-2-one |
| C15H16O2 | |
| MFCD00079518 | |
| nabumetone, relafen, 4-6-methoxynaphthalen-2-yl butan-2-one, arthaxan, nabumetona, relifex, listran, relifen, nabumeton, consolan | |
| CHEBI:7443 | |
| CC(=O)CCC1=CC2=C(C=C1)C=C(C=C2)OC |
Tekniske data
| Nabumetone | |
| C15H16O2 | |
| 25g | |
| nabumetone, relafen, 4-6-methoxynaphthalen-2-yl butan-2-one, arthaxan, nabumetona, relifex, listran, relifen, nabumeton, consolan | |
| BLXXJMDCKKHMKV-UHFFFAOYSA-N | |
| 4-(6-methoxynaphthalen-2-yl)butan-2-one | |
| 4409 | |
| 228.29 |
| 42924-53-8 | |
| MFCD00079518 | |
| 14,6343 | |
| Soluble in alcohol or chloroform | |
| CC(=O)CCC1=CC2=C(C=C1)C=C(C=C2)OC | |
| 228.291 | |
| CHEBI:7443 |