Få mere at vide
N2,N5-Dibenzoyl-4-oxo-L-ornithine Methyl Ester, TRC
Artikelnummer. 30356266
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356266 250 mg
30367955 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356266 Leverandør Toronto Research Chemicals Leverandørnr. TRCD417315250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78420-25-4
Molekylær formel C20H20N2O5
Molekylvægt (g/mol) 368.38
Synonym 2,5-Di(benzoylamino)-4-oxo-pentanoic Acid Methyl Ester
SMIL COC(=O)[C@H](CC(=O)CNC(=O)c1ccccc1)NC(=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale N2,N5-Dibenzoyl-4-oxo-L-ornithine Methyl Ester
Mængde 250 mg
InChI formel InChI=1S/C20H20N2O5/c1-27-20(26)17(22-19(25)15-10-6-3-7-11-15)12-16(23)13-21-18(24)14-8-4-2-5-9-14/h2-11,17H,12-13H2,1H3,(H,21,24)(H,22,25)/t17-/m0/s1
Formel vægt 368.1372