Få mere at vide
N-(Phenylmethyl)-D-tryptophan, TRC
Artikelnummer. 30415360
Skift visning
Mængde:
500 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30415360 2.5 g 2.5g
30422366 1 g -
30415380 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30415360 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9472102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 888009-83-4
Molekylær formel C18H18N2O2
Molekylvægt (g/mol) 294.35
IUPAC navn (2R)-2-(benzylamino)-3-(1H-indol-3-yl)propanoic acid
SMIL OC(=O)[C@@H](Cc1c[nH]c2ccccc12)NCc3ccccc3
Tekniske data
Kemisk navn eller materiale N-(Phenylmethyl)-D-tryptophan
InChI formel InChI=1S/C18H18N2O2/c21-18(22)17(19-11-13-6-2-1-3-7-13)10-14-12-20-16-9-5-4-8-15(14)16/h1-9,12,17,19-20H,10-11H2,(H,21,22)/t17-/m1/s1
Mængde 2.5 g
Formel vægt 294.1368