Få mere at vide
N-Octylpyridin-4-amine, TRC
Artikelnummer. 30132409
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132409 25 g
30003420 1 g
30009618 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132409 Leverandør Toronto Research Chemicals Leverandørnr. TRCO29395025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64690-19-3
Molekylær formel C13 H22 N2
Molekylvægt (g/mol) 206.33
Synonym N-Octyl-4-pyridinamine, 4-(Octylamino)pyridine;
IUPAC navn N-octylpyridin-4-amine
SMIL CCCCCCCCNc1ccncc1
Tekniske data
Kemisk navn eller materiale N-Octylpyridin-4-amine
Renhedsgrad noter GC
Mængde 25 g
InChI formel InChI=1S/C13H22N2/c1-2-3-4-5-6-7-10-15-13-8-11-14-12-9-13/h8-9,11-12H,2-7,10H2,1H3,(H,14,15)
Formel vægt 206.1783
Procent renhed >95%