Få mere at vide
N-Nitrosopiperazine, TRC
Artikelnummer. 17963815
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963815 100 mg
17954925 50 mg
17960685 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17963815 Leverandør Toronto Research Chemicals Leverandørnr. TRCN545350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5632-47-3
Molekylær formel C4 H9 N3 O
Molekylvægt (g/mol) 115.13
Synonym Piperazine, 1-nitroso-, 1-Nitrosopiperazine, N-Mononitrosopiperazine, N-Nitrosopiperazine, NSC 50269, NSC 525340
IUPAC navn 1-nitrosopiperazine
SMIL O=NN1CCNCC1
Tekniske data
Kemisk navn eller materiale N-Nitrosopiperazine
Mængde 100 mg
InChI formel InChI=1S/C4H9N3O/c8-6-7-3-1-5-2-4-7/h5H,1-4H2
Formel vægt 115.0746