Få mere at vide
N-Nitrosocytisine, TRC
Artikelnummer. 30008051
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008051 25 mg
30009160 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008051 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49376525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19634-60-7
Molekylær formel C11 H13 N3 O2
Molekylvægt (g/mol) 219.24
Synonym (1R,5R)-1,2,3,4,5,6-Hexahydro-3-nitroso-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, (1R)-1,2,3,4,5,6-Hexahydro-3-nitroso-1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 12-nitrosocytisine, Nitrosocytisine
SMIL O=NN1C[C@H]2C[C@@H](C1)C3=CC=CC(=O)N3C2
Tekniske data
Kemisk navn eller materiale N-Nitrosocytisine
Mængde 25 mg
InChI formel InChI=1S/C11H13N3O2/c15-11-3-1-2-10-9-4-8(6-14(10)11)5-13(7-9)12-16/h1-3,8-9H,4-7H2/t8-,9+/m1/s1
Formel vægt 219.1008