Få mere at vide
N,N-Dimethyl-3-(p-trifluoromethylphenoxy)-3-phenylpropylamine, TRC
Artikelnummer. 30008311
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008311 1 g
30009042 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008311 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1784801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56225-81-1
Molekylær formel C18H20F3NO
Molekylvægt (g/mol) 323.35
Synonym N,N-Dimethyl-gamma-[4-(trifluoromethyl)phenoxy]benzenepropanamine, N,N-Dimethyl-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]amine;N,N-Dimethyl-3-phenyl-3-(4-(trifluoromethyl)phenoxy)propan-1-amine
IUPAC navn N,N-dimethyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine
SMIL CN(C)CCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-3-(p-trifluoromethylphenoxy)-3-phenylpropylamine
Mængde 1 g
InChI formel InChI=1S/C18H20F3NO/c1-22(2)13-12-17(14-6-4-3-5-7-14)23-16-10-8-15(9-11-16)18(19,20)21/h3-11,17H,12-13H2,1-2H3