Få mere at vide
N,N’-Di-Boc-(L)-cystine-dimethyl Ester, TRC
Artikelnummer. 30378472
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378472 10 g
30378815 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378472 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42200010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 77826-55-2
Molekylær formel C18H32N2O8S2
Molekylvægt (g/mol) 468.59
Synonym N,N’-Di-Boc-(L)-Cystine-Dimethyl Ester
SMIL COC(=O)[C@H](CSSC[C@H](NC(=O)OC(C)(C)C)C(=O)OC)NC(=O)OC(C)(C)C
Tekniske data
Kemisk navn eller materiale N,N'-Di-Boc-(L)-cystine-dimethyl Ester
InChI formel InChI=1S/C18H32N2O8S2/c1-17(2,3)27-15(23)19-11(13(21)25-7)9-29-30-10-12(14(22)26-8)20-16(24)28-18(4,5)6/h11-12H,9-10H2,1-8H3,(H,19,23)(H,20,24)/t11-,12-/m0/s1
Mængde 10 g