Få mere at vide
(-)-N,N-Bisdesmethyl Tramadol, TRC
Artikelnummer. 30137341
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137341 25 mg
30005638 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137341 Leverandør Toronto Research Chemicals Leverandørnr. TRCB42610025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 185502-39-0
Molekylær formel C14H21NO2
Molekylvægt (g/mol) 235.32
Synonym (1S,2S)-2-(Aminomethyl)-1-(3-methoxyphenyl)-cyclohexanol;
IUPAC navn (1R,2R)-2-(aminomethyl)-1-(3-methoxyphenyl)cyclohexan-1-ol
SMIL O[C@@]1(C2=CC(OC)=CC=C2)CCCC[C@@]1([H])CN
Tekniske data
Kemisk navn eller materiale (-)-N,N-Bisdesmethyl Tramadol
Mængde 25 mg
InChI formel InChI=1S/C14H21NO2/c1-17-13-7-4-6-11(9-13)14(16)8-3-2-5-12(14)10-15/h4,6-7,9,12,16H,2-3,5,8,10,15H2,1H3/t12-,14+/m1/s1
Formel vægt 235.1572