Få mere at vide
N-Methylnicotinamide, TRC
Artikelnummer. 17911515
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17911515 5 g -
17900715 2.5 g 2.5g
17901515 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17911515 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3211705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114-33-0
Molekylær formel C7 H8 N2 O
Molekylvægt (g/mol) 136.15
Synonym 3-Pyridinecarboxamide, N-methyl-, Nicotinamide, N-methyl- (6CI,7CI,8CI), N-Methyl-3-pyridinecarboxamide, 3-(Methylcarbamoyl)pyridine, 3-(N-Methylcarbamoyl)pyridine, N-Methylnicotinamide, NSC 66521, Nicotinic acid methylamide, N'-Methylnicotinamide, SR 4415
IUPAC navn N-methylpyridine-3-carboxamide
SMIL CNC(=O)c1cccnc1
Tekniske data
Kemisk navn eller materiale N-Methylnicotinamide
Mængde 5 g
InChI formel InChI=1S/C7H8N2O/c1-8-7(10)6-3-2-4-9-5-6/h2-5H,1H3,(H,8,10)
Formel vægt 136.0637