CAS RN 114-33-0
IUPAC Navn:
N-methylpyridine-3-carboxamide
Synonymer:
N'methylnicotinamide nicotinyl methylamide
Molekylvægt (g/mol):
136.15
Molekylformel:
C7H8N2O
InChi Key:
ZYVXHFWBYUDDBM-UHFFFAOYSA-N
SMILES:
CNC(=O)C1=CC=CN=C1
1
–
2
af
2
Resultater
1
N-Methylnicotinamide, TRC
CAS: 114-33-0 Molekylær formel: C7 H8 N2 O Molekylvægt (g/mol): 136.15 Synonym: 3-Pyridinecarboxamide, N-methyl-,Nicotinamide, N-methyl- (6CI,7CI,8CI),N-Methyl-3-pyridinecarboxamide,3-(Methylcarbamoyl)pyridine,3-(N-Methylcarbamoyl)pyridine,N-Methylnicotinamide,NSC 66521,Nicotinic acid methylamide,N'-Methylnicotinamide,SR 4415 IUPAC navn: N-methylpyridine-3-carboxamide SMIL: CNC(=O)c1cccnc1
2
N-Methylnicotinamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.