Få mere at vide
n-Methylhexylamine, TRC
Artikelnummer. 30475496
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475496 500 mg
30468578 250 mg
30474513 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475496 Leverandør Toronto Research Chemicals Leverandørnr. TRCM336470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35161-70-7
Molekylær formel C7H17N
Molekylvægt (g/mol) 115.22
Synonym N-Methyl-1-hexanamine, Hexylmethylamine, Methylhexylamine, N-Hexyl-N-methylamine, N-Hexylmethylamine, N-Methyl-1-aminohexane, N-Methyl-1-hexylamine, N-Methyl-N-hexylamine, N-Methylhexanamine
IUPAC navn N-methylhexan-1-amine
SMIL CCCCCCNC
Tekniske data
Kemisk navn eller materiale n-Methylhexylamine
Mængde 500 mg
InChI formel InChI=1S/C7H17N/c1-3-4-5-6-7-8-2/h8H,3-7H2,1-2H3
Formel vægt 115.1361