Få mere at vide
N-Methyldopamine hydrochloride, 95%, Thermo Scientific™
Dopamine agonist
2022.42 DKK - 8432.96 DKK
Kemiske identifikatorer
| CAS | 62-32-8 |
|---|---|
| Molekylær formel | C9H14ClNO2 |
| Molekylvægt (g/mol) | 203.67 |
| MDL nummer | MFCD00035074 |
| InChI nøgle | JCDRZCWRRLKLTB-UHFFFAOYSA-N |
| Synonym | n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride |
| PubChem CID | 86470 |
| IUPAC navn | hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride |
| SMIL | [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1 |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15463099
|
Thermo Scientific Alfa Aesar
J60306.03 |
1g |
2022.42 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15473099
|
Thermo Scientific Alfa Aesar
J60306.06 |
5g |
8432.96 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAndregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine′s.
Solubility
Soluble in methanol, water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Kemiske identifikatorer
| C9H14ClNO2 | |
| MFCD00035074 | |
| n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride | |
| hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride |
Tekniske data
| N-Methyldopamine hydrochloride | |
| C9H14ClNO2 | |
| n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride | |
| JCDRZCWRRLKLTB-UHFFFAOYSA-N | |
| hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride | |
| 86470 | |
| 95% |
| 62-32-8 | |
| MFCD00035074 | |
| Soluble in methanol,water. | |
| [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1 | |
| 203.67 | |
| 203.67 |
Sikkerhed og håndtering
missing translation for 'einecsNumber' : 200-528-3
missing translation for 'tsca' : No
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.