Få mere at vide
N-Methyldopamine hydrochloride, 95%, Thermo Scientific™
Dopamine agonist
Brand:  Thermo Scientific Alfa Aesar J60306.03
Mængde 1g
 Se flere versioner af dette produkt
Produktkode 15463099
  • 2022.42 DKK / 1g
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Andregenic and dopamine receptor agonist; active metabolite of Ibopramine. Also, it is used as internal standard in chromatographic analysis of catecholamine′s.
Solubility
Soluble in methanol, water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemiske identifikatorer
62-32-8
203.67
JCDRZCWRRLKLTB-UHFFFAOYSA-N
86470
[H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1
C9H14ClNO2
MFCD00035074
n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride
hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
Tekniske data
N-Methyldopamine hydrochloride
C9H14ClNO2
1g
Soluble in methanol,water.
[H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1
203.67
203.67
62-32-8
MFCD00035074
n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride
JCDRZCWRRLKLTB-UHFFFAOYSA-N
hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
86470
95%