Få mere at vide
N-Formyl-3-mercaptovaline, TRC
Artikelnummer. 17903445
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17903445 250 mg
17923895 500 mg
17944255 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17903445 Leverandør Toronto Research Chemicals Leverandørnr. TRCF701155250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 158411-82-6
Molekylær formel C6 H11 N O3 S
Molekylvægt (g/mol) 177.22
Synonym N-Formyl-3-mercaptovaline, DL-Valine, N-formyl-3-mercapto-, NSC 28038, N-Formylpenicillamin
IUPAC navn 2-formamido-3-methyl-3-sulfanylbutanoic acid
SMIL CC(C)(S)C(NC=O)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Formylpenicillamin
InChI formel InChI=1S/C6H11NO3S/c1-6(2,11)4(5(9)10)7-3-8/h3-4,11H,1-2H3,(H,7,8)(H,9,10)
Mængde 250 mg
Formel vægt 177.046