Få mere at vide
N-Ethyl-3-buteno-o-toluidide, TRC
Artikelnummer. 30375785
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375785 500 mg
30374856 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30375785 Leverandør Toronto Research Chemicals Leverandørnr. TRCE900630500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13936-71-5
Molekylær formel C13 H17 N O
Molekylvægt (g/mol) 203.28
Synonym N-Ethyl-N-(2-methylphenyl)-3-butenamide, N-Ethyl-3-buteno-o-toluidide (8Cl)
IUPAC navn N-ethyl-N-(2-methylphenyl)but-3-enamide
SMIL CCN(C(=O)CC=C)c1ccccc1C
Tekniske data
Kemisk navn eller materiale N-Ethyl-N-(2-methylphenyl)but-3-enamide
Mængde 500 mg
InChI formel InChI=1S/C13H17NO/c1-4-8-13(15)14(5-2)12-10-7-6-9-11(12)3/h4,6-7,9-10H,1,5,8H2,2-3H3
Formel vægt 203.131