Få mere at vide
N-Desmethyl Zopiclone Hydrochloride, TRC
Artikelnummer. 30011222
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011222 10 mg
30010508 25 mg
30010067 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011222 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29677010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216659-29-8
Molekylær formel C16 H15 Cl N6 O3 . Cl H
Molekylvægt (g/mol) 411.24
Synonym 1-Piperazinecarboxylic acid, 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, hydrochloride (1:1), N-DemethylzopicloneHydrochloride
IUPAC navn [6-(5-chloropyridin-2-yl)-5-oxo-7H-pyrrolo[3,4-b]pyrazin-7-yl] piperazine-1-carboxylate;hydrochloride
SMIL Cl.Clc1ccc(nc1)N2C(OC(=O)N3CCNCC3)c4nccnc4C2=O
Tekniske data
Kemisk navn eller materiale N-Demethylzopiclone hydrochloride
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C16H15ClN6O3.ClH/c17-10-1-2-11(21-9-10)23-14(24)12-13(20-4-3-19-12)15(23)26-16(25)22-7-5-18-6-8-22;/h1-4,9,15,18H,5-8H2;1H
Formel vægt 410.0661
Procent renhed >95%