Få mere at vide
N-Desmethyl Droloxifene (contains up to 5% Z isomer), TRC
Artikelnummer. 30367601
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367601 10 mg
30359888 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367601 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29202510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83647-33-0
Molekylær formel C25 H27 N O2
Molekylvægt (g/mol) 373.49
IUPAC navn 3-[(Z)-2-[4-[2-(methylamino)ethoxy]phenyl]-1-phenylbut-1-enyl]phenol
SMIL CC\C(=C(/c1ccccc1)\c2cccc(O)c2)\c3ccc(OCCNC)cc3
Tekniske data
Kemisk navn eller materiale N-Desmethyl Droloxifene
Mængde 10 mg
InChI formel InChI=1S/C25H27NO2/c1-3-24(19-12-14-23(15-13-19)28-17-16-26-2)25(20-8-5-4-6-9-20)21-10-7-11-22(27)18-21/h4-15,18,26-27H,3,16-17H2,1-2H3/b25-24-
Formel vægt 373.2042