Få mere at vide
N-Desmethyl Diltiazem-d4 Hydrochloride, TRC
Artikelnummer. 30362211
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362211 10 mg
30355762 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362211 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29161310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217650-51-5
Molekylær formel C21H21D4ClN2O4S
Molekylvægt (g/mol) 440.98
Synonym (2S-cis)-3-(Acetyloxy)-2,3-dihydro-2-(4-methoxyphenyl)-5-[2-(methylamino-d4)ethyl]-1,5-benzothiazepin-4(5H)-one Monohydrochloride, N-Demethyldiltiazem-d4, N-Monodesmethyldiltiazem-d4
SMIL O=C(N(C([2H])([2H])C([2H])([2H])NC)C(C=CC=C1)=C1S2)[C@H](OC(C)=O)[C@@H]2C3=CC=C(OC)C=C3.Cl
Tekniske data
Kemisk navn eller materiale N-Desmethyl Diltiazem-d4 Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C21H24N2O4S.ClH/c1-14(24)27-19-20(15-8-10-16(26-3)11-9-15)28-18-7-5-4-6-17(18)23(21(19)25)13-12-22-2;/h4-11,19-20,22H,12-13H2,1-3H3;1H/t19-,20+;/m1./s1/i12D2,13D2;