Få mere at vide
N-Demethyl Trimebutine-d5 Hydrochloride, TRC
Artikelnummer. 30365610
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365610 1 mg
30354767 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365610 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2313421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1286632-72-1
Molekylær formel C21 H22 D5 N O5 . H Cl
Molekylvægt (g/mol) 414.93
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])C(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NC)c2ccccc2
Tekniske data
Kemisk navn eller materiale N-Demethyl Trimebutine-d5 hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C21H27NO5.ClH/c1-6-21(22-2,16-10-8-7-9-11-16)14-27-20(23)15-12-17(24-3)19(26-5)18(13-15)25-4;/h7-13,22H,6,14H2,1-5H3;1H/i1D3,6D2;