Få mere at vide
N-D-Gluconoyl-L-methionine, TRC
Artikelnummer. 30240536
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240536 5 mg
30240583 25 mg
30240779 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240536 Leverandør Toronto Research Chemicals Leverandørnr. TRCG9423105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 94231-87-5
Molekylær formel C11H21NO8S
Molekylvægt (g/mol) 327.35
Synonym (S)-4-(Methylthio)-2-((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanamido)butanoic Acid;
IUPAC navn (2S)-4-methylsulfanyl-2-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]butanoic acid
SMIL CSCC[C@H](NC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)C(=O)O
Tekniske data
Kemisk navn eller materiale N-D-Gluconoyl-L-methionine
InChI formel InChI=1S/C11H21NO8S/c1-21-3-2-5(11(19)20)12-10(18)9(17)8(16)7(15)6(14)4-13/h5-9,13-17H,2-4H2,1H3,(H,12,18)(H,19,20)/t5-,6+,7+,8-,9+/m0/s1
Mængde 5 mg
Formel vægt 327.0988