Få mere at vide
N’-Cyano-N,N-dimethylguanidine, TRC
Artikelnummer. 30367453
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367453 10 mg
30363656 100 mg
30350382 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367453 Leverandør Toronto Research Chemicals Leverandørnr. TRCC97924010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1609-06-9
Molekylær formel C4 H8 N4
Molekylvægt (g/mol) 112.13
Synonym 1,1-Dimethyl-3-cyanoguanidine
IUPAC navn 3-cyano-1,1-dimethylguanidine
SMIL CN(C)C(=N)NC#N
Tekniske data
Kemisk navn eller materiale 1,1-Dimethyl-3-cyanoguanidine
Mængde 10 mg
InChI formel InChI=1S/C4H8N4/c1-8(2)4(6)7-3-5/h1-2H3,(H2,6,7)
Formel vægt 112.0749