missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophanol, 98% Product Code.: 15423777

Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophanol, 98%

Product Code. p-7030211
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15423777

Brand: Thermo Scientific Chemicals H27763.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158932-00-4
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.36
MDL Number MFCD00270221
InChI Key JEFQUFUAEKORKL-GFCCVEGCSA-N
Synonym boc-d-tryptophanol, n-boc-d-tryptophanol, r-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, n alpha-boc-d-tryptophanol, boc-d-trp-ol, tert-butyl n-2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, ambotzbal1034, n-t-boc-d-tryptophanol, tert-butyl r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, tert-butyl 2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate
PubChem CID 20744818
SMILES CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 119°C to 121°C
Quantity 250 mg
Optical Rotation +53° (c=2 in Acetic Acid)
IUPAC Name tert-butyl N-[1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Formula Weight 290.36
Percent Purity 98%
Chemical Name or Material N(alpha)-Boc-D-tryptophanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot Number10148234 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH27763.06,H27763.MD,H27763.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophanol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.