Få mere at vide
N-Acetyl Serotonin Beta-D-Glucuronide, TRC
Artikelnummer. 30135318
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135318 5 mg
30007832 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135318 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1877905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18430-06-3
Molekylær formel C18 H22 N2 O8
Molekylvægt (g/mol) 394.38
Synonym N-Acetyl Serotonin-B-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[[3-(2-acetamidoethyl)-1H-indol-5-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL CC(=O)NCCc1c[nH]c2ccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)cc12
Tekniske data
Kemisk navn eller materiale N-Acetyl Serotonin beta-D-Glucuronide
Mængde 5 mg
InChI formel InChI=1S/C18H22N2O8/c1-8(21)19-5-4-9-7-20-12-3-2-10(6-11(9)12)27-18-15(24)13(22)14(23)16(28-18)17(25)26/h2-3,6-7,13-16,18,20,22-24H,4-5H2,1H3,(H,19,21)(H,25,26)/t13-,14-,15+,16-,18+/m0/s1
Formel vægt 394.1376